Learn More
Aviva Systems Biology Corp ZNF312 Antibody - N-terminal region (ARP33141_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33141_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF312. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF312. Alternate names: FEZ, TOF, FEZL, FKSG36, ZFP312, ZNF312ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-52893