Learn More
Aviva Systems Biology Corp ZNF313 Antibody - C-terminal region (ARP39375_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39375_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF313. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF313. Alternate names: PSORS12, ZNF313ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-284-801