Learn More
Aviva Systems Biology Corp ZNF319 Antibody - N-terminal region (ARP50648_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50648_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF319. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF319. Alternate names: MGC126816, ZFP319ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-60400