Learn More
Aviva Systems Biology Corp ZNF320 Antibody - N-terminal region (ARP50901_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50901_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF320. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF320. Alternate names: DKFZp686G16228, ZFPLApplicationWB. Species reactivity: Human
Catalog No. 50-285-550