Learn More
Aviva Systems Biology Corp ZNF323 Antibody - middle region (ARP36063_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36063_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF323. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF323. Alternate names: ZNF310P, ZNF20-Lp, dJ874C20.2, ZNF323ApplicationWB. Species reactivity: Human
Catalog No. 50-014-55997