Learn More
Aviva Systems Biology Corp ZNF326 Antibody - C-terminal region (ARP40068_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40068_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF326. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF326. Alternate names: FLJ20403, MGC61591, ZAN75, Zfp326, dJ871E2.1ApplicationIHC, WB. Species reactivity: Dog, Human, Rat
Catalog No. 50-014-53172