Learn More
Aviva Systems Biology Corp ZNF326 Antibody - C-terminal region (ARP31894_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31894_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF326. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF326. Alternate names: FLJ20403, MGC61591, ZAN75, Zfp326, dJ871E2.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54355