Learn More
Aviva Systems Biology Corp ZNF336 Antibody - N-terminal region (ARP33281_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33281_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF336. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF336. Alternate names: ZBTB23, ZNF336ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54942