Learn More
Aviva Systems Biology Corp ZNF337 Antibody - middle region (ARP39204_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39204_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF337. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF337. Alternate names: -ApplicationWB. Species reactivity: Human
Catalog No. 50-014-56940