Learn More
Aviva Systems Biology Corp ZNF33A Antibody - middle region (ARP51610_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51610_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF33A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF33A. Alternate names: FLJ23404, KIAA0065, KOX2, KOX31, KOX5, NF11A, ZNF11, ZNF11A, ZNF33, ZZAPKApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-53660