Learn More
Aviva Systems Biology Corp ZNF342 Antibody - middle region (ARP32196_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32196_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF342. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF342. Alternate names: ZNF296, ZFP296, ZNF342ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54484