Learn More
Aviva Systems Biology Corp ZNF350 Antibody - N-terminal region (ARP37781_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37781_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF350. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF350. Alternate names: ZBRK1, ZFQRApplicationWB. Species reactivity: Dog, Horse, Human
Catalog No. 50-014-56430