Learn More
Aviva Systems Biology Corp ZNF354A Antibody - middle region (ARP33415_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33415_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF354A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF354A. Alternate names: EZNF, HKL1, KID-1, KID1, TCF17ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55011