Learn More
Aviva Systems Biology Corp ZNF358 Antibody - N-terminal region (ARP35948_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35948_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF358. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF358. Alternate names: ZFENDApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-55938