Learn More
Aviva Systems Biology Corp ZNF358 Antibody - C-terminal region (ARP57957_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57957_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF358. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF358. Alternate names: FLJ10390, ZFENDApplicationWB. Species reactivity: Cow, Human
Catalog No. 50-014-62192