Learn More
Aviva Systems Biology Corp ZNF364 Antibody - C-terminal region (ARP43154_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43154_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF364. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF364. Alternate names: BCA2, RNF115, ZNF364ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53358