Learn More
Aviva Systems Biology Corp ZNF366 Antibody - C-terminal region (ARP39922_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39922_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF366. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF366. Alternate names: FLJ39796, DCSCRIPTApplicationWB. Species reactivity: Guinea Pig, Human, Pig, Rabbit
Catalog No. 50-014-57233