Learn More
Aviva Systems Biology Corp ZNF37A Antibody - middle region (ARP47499_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47499_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF37A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ZNF37A. Alternate names: FLJ3472, KOX21, ZNF37ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-310