Learn More
Aviva Systems Biology Corp ZNF384 Antibody - middle region (ARP31601_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31601_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF384. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF384. Alternate names: CAGH1, CAGH1A, CIZ, ERDA2, NMP4, NP, TNRC1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54277