Learn More
Aviva Systems Biology Corp ZNF385 Antibody - C-terminal region (ARP31849_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31849_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF385. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF385. Alternate names: HZF, RZF, ZFP385, ZNF385ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54339