Learn More
Aviva Systems Biology Corp ZNF391 Antibody - N-terminal region (ARP50450_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50450_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF391. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF391. Alternate names: dJ153G14.3ApplicationWB. Species reactivity: Cow, Dog, Human
Catalog No. 50-014-60347