Learn More
Aviva Systems Biology Corp ZNF395 Antibody - C-terminal region (ARP33162_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33162_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF395. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF395. Alternate names: PBF, PRF1, HDBP2, PRF-1, HDBP-2, HDRF-2, Si-1-8-14ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-52896