Learn More
Aviva Systems Biology Corp ZNF396 Antibody - middle region (ARP34807_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34807_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF396. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF396. Alternate names: FLJ31213, ZSCAN14ApplicationWB. Species reactivity: Human
Catalog No. 50-014-55571