Learn More
Aviva Systems Biology Corp ZNF397 Antibody - C-terminal region (ARP39691_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39691_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF397. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF397. Alternate names: ZNF47, ZSCAN15ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-57139