Learn More
Aviva Systems Biology Corp ZNF398 Antibody - middle region (ARP32953_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32953_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF398. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF398. Alternate names: KIAA1339, P51, P71, ZER6ApplicationWB. Species reactivity: Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54830