Learn More
Aviva Systems Biology Corp ZNF408 Antibody - middle region (ARP33594_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33594_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF408. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF408. Alternate names: FLJ12827ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Zebrafish
Catalog No. 50-014-55084