Learn More
Aviva Systems Biology Corp ZNF414 Antibody - N-terminal region (ARP39692_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39692_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF414. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF414. Alternate names: MGC15716, Zfp414, ZFP414ApplicationWB. Species reactivity: Dog, Horse, Human
Catalog No. 50-014-57140