Learn More
Aviva Systems Biology Corp ZNF415 Antibody - C-terminal region (ARP39352_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39352_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF415. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF415. Alternate names: FLJ11191ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-53136