Learn More
Aviva Systems Biology Corp ZNF419 Antibody - N-terminal region (ARP36041_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36041_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF419. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF419. Alternate names: FLJ23233, ZNF419AApplicationWB. Species reactivity: Human
Catalog No. 50-014-55984