Learn More
Aviva Systems Biology Corp ZNF426 Antibody - N-terminal region (ARP33566_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33566_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF426. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF426. Alternate names: ApplicationWB. Species reactivity: Human, Mouse, Rat
Catalog No. 50-014-55075