Learn More
Aviva Systems Biology Corp ZNF43 Antibody - middle region (ARP31980_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31980_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF43. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF43. Alternate names: DKFZp686L1854, HTF6, KOX27, ZNF39L1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Yeast, Zebrafish
Catalog No. 50-014-54401