Learn More
Aviva Systems Biology Corp ZNF431 Antibody - middle region (ARP39802_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39802_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF431. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF431. Alternate names: DKFZp313E0830, KIAA1969ApplicationWB. Species reactivity: Human
Catalog No. 50-014-57183