Learn More
Aviva Systems Biology Corp ZNF433 Antibody - N-terminal region (ARP36196_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36196_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF433. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF433. Alternate names: FLJ40981ApplicationWB. Species reactivity: Dog, Human, Pig, Zebrafish
Catalog No. 50-014-56061