Learn More
Aviva Systems Biology Corp ZNF434 Antibody - N-terminal region (ARP35939_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35939_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF434. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF434. Alternate names: FLJ20417, FLJ31901, MGC4179, ZNF434ApplicationWB. Species reactivity: Human
Catalog No. 50-014-55934