Learn More
Aviva Systems Biology Corp ZNF440 Antibody - N-terminal region (ARP39886_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39886_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF440. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF440. Alternate names: ApplicationWB. Species reactivity: Human
Catalog No. 50-014-57219