Learn More
Aviva Systems Biology Corp ZNF440 Antibody - middle region (ARP36174_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36174_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF440. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF440. Alternate names: FLJ37933ApplicationWB. Species reactivity: Human
Catalog No. 50-014-56049