Learn More
Aviva Systems Biology Corp ZNF441 Antibody - N-terminal region (ARP37815_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37815_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF441. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF441. Alternate names: ApplicationWB. Species reactivity: Human
Catalog No. 50-014-56441