Learn More
Aviva Systems Biology Corp ZNF444 Antibody - C-terminal region (ARP39348_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39348_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF444. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF444. Alternate names: EZF2, EZF-2, ZSCAN17ApplicationIHC, WB. Species reactivity: Cow, Dog, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56997