Learn More
Aviva Systems Biology Corp ZNF444 Antibody - middle region (ARP35954_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35954_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF444. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF444. Alternate names: EZF-2, EZF2, FLJ11137, ZSCAN17ApplicationWB. Species reactivity: Human
Catalog No. 50-014-55943