Learn More
Aviva Systems Biology Corp ZNF446 Antibody - N-terminal region (ARP39318_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39318_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF446. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF446. Alternate names: FLJ20626, ZKSCAN20, ZSCAN30ApplicationWB. Species reactivity: Human, Mouse, Rat
Catalog No. 50-014-56983