Learn More
Aviva Systems Biology Corp ZNF447 Antibody - N-terminal region (ARP33305_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33305_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF447. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF447. Alternate names: ZNF447ApplicationWB. Species reactivity: Dog, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54953