Learn More
Aviva Systems Biology Corp ZNF451 Antibody - C-terminal region (ARP39198_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39198_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF451. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF451. Alternate names: COASTER, FLJ90693, KIAA0576, MGC26701, dJ417I1.1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56938