Learn More
Aviva Systems Biology Corp ZNF471 Antibody - N-terminal region (ARP35981_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35981_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF471. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF471. Alternate names: ERP1, KIAA1396, MGC150657, MGC150658, Z1971ApplicationWB. Species reactivity: Horse, Human, Mouse, Pig
Catalog No. 50-014-55953