Learn More
Aviva Systems Biology Corp ZNF474 Antibody - N-terminal region (ARP32163_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32163_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF474. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF474. Alternate names: FLJ32921ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-54469