Learn More
Aviva Systems Biology Corp ZNF485 Antibody - N-terminal region (ARP39861_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39861_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF485. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF485. Alternate names: -ApplicationWB. Species reactivity: Dog, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53157