Learn More
Aviva Systems Biology Corp ZNF486 Antibody - middle region (ARP51696_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51696_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF486. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF486. Alternate names: KRBO2, MGC2396ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60589