Learn More
Aviva Systems Biology Corp ZNF488 Antibody - middle region (ARP36211_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36211_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF488. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF488. Alternate names: FLJ32104ApplicationWB. Species reactivity: Human
Catalog No. 50-014-56069