Learn More
Aviva Systems Biology Corp ZNF491 Antibody - N-terminal region (ARP36172_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36172_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF491. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF491. Alternate names: FLJ34791, MGC126639, MGC126641ApplicationWB. Species reactivity: Human
Catalog No. 50-014-56047