Learn More
Aviva Systems Biology Corp ZNF496 Antibody - middle region (ARP30024_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30024_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF496. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF496. Alternate names: MGC15548, NIZP1, ZKSCAN17, ZFP496ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-54103