Learn More
Aviva Systems Biology Corp ZNF498 Antibody - C-terminal region (ARP39846_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39846_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF498. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF498. Alternate names: ZSCAN25, ZNF498ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57200